Learn More
Med Vet International Mathieu Needle Holder, 6.25-in, Spring Handles, Serrated Jaws, Von Klaus German Surgical Steel
Catalog No. 502092499
Encompass_Preferred
Mathieu needle holders have a plier handle grip with spring handles, serrated jaws, and a curved base that can lock and unlock. Mathieu needle holders are used when a more refined suturing is needed. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2499 Supplier Med Vet International Supplier No. VK1112017
Mathieu needle holders have a plier handle grip with spring handles, serrated jaws, and a curved base that can lock and unlock. Mathieu needle holders are used when a more refined suturing is needed. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.