Learn More
Med Vet International Miltex Castroviejo Needle Holder, Round Handle w/Lock, 4.87in, German, 18-1816Tc
Catalog No. 502829148
Encompass_Preferred
With needle holder jaws made of tungsten carbide, the hardest steel used in surgical instruments, this instrument ensures prolonged durability and exceptional quality. The edges around the box lock have distinct non-catch bevel. 4.87in total length. Made with German steel.
Catalog No. 50-282-9148 Supplier Med Vet International Supplier No. MIL181816TC
With needle holder jaws made of tungsten carbide, the hardest steel used in surgical instruments, this instrument ensures prolonged durability and exceptional quality. The edges around the box lock have distinct non-catch bevel. 4.87in total length. Made with German steel.