Learn More
Med Vet International Miltex Meisterhand Mayo-Hegar Masson Needle Holder, Serrated Jaws, Tungsten, 6.25in, German, Mh8-44Tc
Catalog No. 502830967
Encompass_Preferred
Versatile in it’s design, Mayo-Hegar needle holders are frequently used in both human and animal procedures. MeisterHand build provides excellent craftsmanship. Tungsten carbide provides increased protection against corrosion and abrasions. Serrated jaws to allow greater grip on items held. Tungsten carbide coating helps prevent corrosion and abrasions. 6.25in total length. Made with top quality German steel.
Catalog No. 50-283-0967 Supplier Med Vet International Supplier No. MILMH844TC
Versatile in it’s design, Mayo-Hegar needle holders are frequently used in both human and animal procedures. MeisterHand build provides excellent craftsmanship. Tungsten carbide provides increased protection against corrosion and abrasions. Serrated jaws to allow greater grip on items held. Tungsten carbide coating helps prevent corrosion and abrasions. 6.25in total length. Made with top quality German steel.