Learn More
Med Vet International Dynarex Securesafe Safety Hypodermic Needle, 25g, 1 1/2" Needle, 1000/Case
Catalog No. 502831489
Encompass_Preferred
The Dynarex SecureSafe Safety Hypodermic Needles provide a sterile, effective way to deliver comfortable injections. All needles are color coded to represent the gauge of the needle are compatible with both luer lock and luer slip syringes. The thin wall needles have a double bevel, allowing for ease of penetration while helping larger doses flow smoothly. The SecureSafe safety mechanism features a quick cover that enables the user to cover and protect the needle with a press and snap. The Dynarex SecureSafe Safety Hypodermic Needles are designed with a safety mechanism to provide a comfortable, effective way to deliver medication or draw blood, while protecting users from accidental needlesticks.
Catalog No. 50-283-1489 Supplier Med Vet International Supplier No. 8973
The Dynarex SecureSafe Safety Hypodermic Needles provide a sterile, effective way to deliver comfortable injections. All needles are color coded to represent the gauge of the needle are compatible with both luer lock and luer slip syringes. The thin wall needles have a double bevel, allowing for ease of penetration while helping larger doses flow smoothly. The SecureSafe safety mechanism features a quick cover that enables the user to cover and protect the needle with a press and snap. The Dynarex SecureSafe Safety Hypodermic Needles are designed with a safety mechanism to provide a comfortable, effective way to deliver medication or draw blood, while protecting users from accidental needlesticks.