Learn More
Med Vet International Dynarex Hypodermic Needle, Non-Safety, 25g, 5/8" Needle, 1000/Case
Catalog No. 502830215
Encompass_Preferred
The Dynarex Hypodermic Needles are designed to provide a comfortable, effective way to deliver medication or draw blood. Color-coded needle hub for quick identification of the needle gauge Compatible with luer lock and luer slip syringes Non-Toxic Non-Pyrogenic Not made with natural rubber latex
Catalog No. 50-283-0215 Supplier Med Vet International Supplier No. 6972
The Dynarex Hypodermic Needles are designed to provide a comfortable, effective way to deliver medication or draw blood. Color-coded needle hub for quick identification of the needle gauge Compatible with luer lock and luer slip syringes Non-Toxic Non-Pyrogenic Not made with natural rubber latex