Learn More
Med Vet International Safety Hypodermic Needles, 25g X 1.5in, 100/Box
Catalog No. 502830340
Encompass_Preferred
Safety Hypodermic Needles, 25G X 1.5in, 100/Box Safety Needle Features One handed activation No solid surface required Simple finger stroke activation Protective arm keeps fingers behind the needle An audible "click" as well as tactile feel and a visual indicator confirm safety mechanism engagement Needle remains locked inside activated cover Fits any brand Syringe
Catalog No. 50-283-0340 Supplier Med Vet International Supplier No. 27406
Safety Hypodermic Needles, 25G X 1.5in, 100/Box Safety Needle Features One handed activation No solid surface required Simple finger stroke activation Protective arm keeps fingers behind the needle An audible "click" as well as tactile feel and a visual indicator confirm safety mechanism engagement Needle remains locked inside activated cover Fits any brand Syringe