Learn More
Trajan Scientific Needle Kit Repair Medical 1/EA
SDP
Supplier:  Trajan Scientific 033630
Encompass_Preferred
Replacemenet needle and plunger for 0 5 L NanoVolume syringe with 7 cm 0 47 mm OD on-column needle
Catalog No. 50-004-94649