Learn More
Trajan Scientific Needle Kit Repair Medical 1/EA
SDP
Catalog No. 5000494711
Encompass_Preferred
Replacemenet needle and plunger for 0 5 L PerkinElmer NanoVolume syringe with 23-gauge needle
Catalog No. 50-004-94711 Supplier Trajan Scientific Supplier No. 033765
Replacemenet needle and plunger for 0 5 L PerkinElmer NanoVolume syringe with 23-gauge needle