Learn More
Trajan Scientific Needle Medical Life Science Lab 1/EA
SDP
Supplier:  Trajan Scientific 038815
Encompass_Preferred
5 cm 0 63 mm OD side hole dome tipped needle for 25 L-2 5 mL syringe valve 031905
Catalog No. 50-004-93942