Learn More
Trajan Scientific Needle Medical Life Science Lab 2/PK
SDP
Catalog No. 5000494732
Encompass_Preferred
4 2 cm 0 63 mm OD cone tipped needle for 5 L Agilent syringes PK2
Catalog No. 50-004-94732 Supplier Trajan Scientific Supplier No. 036720
4 2 cm 0 63 mm OD cone tipped needle for 5 L Agilent syringes PK2