Learn More
Trajan Scientific Needle Medical Life Science Lab 2/PK
SDP
Catalog No. 5000494775
Encompass_Preferred
7 cm 0 63 mm OD 0 37 mm ID side hole needle for high pressure syringes
Catalog No. 50-004-94775 Supplier Trajan Scientific Supplier No. 0315722
7 cm 0 63 mm OD 0 37 mm ID side hole needle for high pressure syringes