Learn More
Trajan Scientific Needle Medical Life Science Lab 2/PK
SDP
Catalog No. 5000494432
Encompass_Preferred
7 5 cm 0 23 mm OD needle for 10 L on-column syringes PK2
Catalog No. 50-004-94432 Supplier Trajan Scientific Supplier No. 037675
7 5 cm 0 23 mm OD needle for 10 L on-column syringes PK2