Learn More
Trajan Scientific Needle Medical Life Science Lab 2/PK
SDP
Catalog No. 5000494001
Encompass_Preferred
5 cm 0 63 mm OD cone tipped needle for 10 L CTC Analytics and Thermo Scientific syringes PK2
Catalog No. 50-004-94001 Supplier Trajan Scientific Supplier No. 037787
5 cm 0 63 mm OD cone tipped needle for 10 L CTC Analytics and Thermo Scientific syringes PK2