Learn More
Trajan Scientific Needle Medical Life Science Lab 2/PK
SDP
Catalog No. 5000494718
Encompass_Preferred
5 cm 0 50 mm OD bevel tipped needle for 10 L Varian 8200 series syringes PK2
Catalog No. 50-004-94718 Supplier Trajan Scientific Supplier No. 037776
5 cm 0 50 mm OD bevel tipped needle for 10 L Varian 8200 series syringes PK2