Learn More
Trajan Scientific Needle Medical Life Science Lab 2/PK
SDP
Supplier:  Trajan Scientific 038736
Encompass_Preferred
5 cm 0 63 mm OD side hole dome tipped needle for 25-500 L CTC Analytics and Thermo Scientific syringes PK2
Catalog No. 50-004-93835