Learn More
Trajan Scientific Needle Medical Life Science Lab 2/PK
SDP
Supplier:  Trajan Scientific 037779
Encompass_Preferred
5 3 cm 0 63 mm OD side hole dome tipped needle for 10 L Varian 8200 series syringes PK2
Catalog No. 50-004-94869