Learn More
Trajan Scientific Needle Medical Life Science Lab 2/PK
SDP
Catalog No. 5000494323
Encompass_Preferred
10 cm 0 17 mm OD vitreous silica needle for 10 L on-column syringes PK2
Catalog No. 50-004-94323 Supplier Trajan Scientific Supplier No. 037610
10 cm 0 17 mm OD vitreous silica needle for 10 L on-column syringes PK2