Learn More
Trajan Scientific Needle Medical Life Science Lab 2/PK
SDP
Catalog No. 5000494844
Encompass_Preferred
11 5 cm 0 50 mm OD cone tipped needle for 25-500 L syringes PK2
Catalog No. 50-004-94844 Supplier Trajan Scientific Supplier No. 038060
11 5 cm 0 50 mm OD cone tipped needle for 25-500 L syringes PK2