Learn More
Trajan Scientific Needle Medical Life Science Lab 2/PK
SDP
Supplier:  Trajan Scientific 0315721
Encompass_Preferred
7 5 cm 0 63 mm OD 0 37 mm ID side hole needle for high pressure syringes
Catalog No. 50-004-94830