Learn More
Trajan Scientific Needle Medical Life Science Lab 2/PK
SDP
Catalog No. 5000494977
Encompass_Preferred
4 2 cm 0 47 mm OD cone tipped needle for 10 L Agilent syringes PK2
Catalog No. 50-004-94977 Supplier Trajan Scientific Supplier No. 037715
4 2 cm 0 47 mm OD cone tipped needle for 10 L Agilent syringes PK2