Learn More
Trajan Scientific Needle Medical Life Science Lab 5/PK
SDP
Catalog No. 5000494825
Encompass_Preferred
11 5 cm 0 47 mm OD bevel tipped needle for 10 L syringes PK5
Catalog No. 50-004-94825 Supplier Trajan Scientific Supplier No. 037160
11 5 cm 0 47 mm OD bevel tipped needle for 10 L syringes PK5