Learn More
Trajan Scientific Needle Medical Life Science Lab 5/PK
SDP
Catalog No. 5000494959
Encompass_Preferred
7 cm 0 50 mm OD bevel tipped needle for 25-500 L syringes PK5
Catalog No. 50-004-94959 Supplier Trajan Scientific Supplier No. 038130
7 cm 0 50 mm OD bevel tipped needle for 25-500 L syringes PK5