Learn More
World Precision Instrument Needle Microprobes, High Temperature-29ga, 3cm (MT-29/3HT)
SDP
Supplier:  World Precision Instrument MT29/3HT
Encompass
When precise temperature measurements are required, WPI can provide you with a very accurate monitor and thermocouple microprobes. WPI monitors have both resolution and accuracy of 0.1°C in the 0-50°C range and are traceable to NIST standards, whereas, other competitive electronic thermometers have an accuracy that is usually to 0.5°C or worse. Furthermore, all our type T clinical probes are guaranteed accurate to 0.1°C, due to our stringent wire standards. These are five times more accurate than competitive probes made with regular “Special Limits” wire.
Catalog No. 50-253-8663