Learn More
Trajan Scientific NEEDLE NLL-11.5/19 5/PK
SDP
Catalog No. 502530253
Encompass_Preferred
Trajan Replaceable Luer Lock Needles: Bevel, Length Needle: 115 mm, 4.52 in., Needle Gauge: 19 G, Needle Point Style: Bevel, Diameter Inner: 0.025 in., 0.65 mm, Diameter Outer: 0.042 in., 1 mm, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle
Catalog No. 50-253-0253 Supplier Trajan Scientific Supplier No. 039831
Add to Cart
Add to Cart
Trajan Replaceable Luer Lock Needles: Bevel, Length Needle: 115 mm, 4.52 in., Needle Gauge: 19 G, Needle Point Style: Bevel, Diameter Inner: 0.025 in., 0.65 mm, Diameter Outer: 0.042 in., 1 mm, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle