Learn More
Electron Microscopy Sciences K-kits Needle Pen
SDP
Catalog No. 501928046
Encompass_Preferred
The Needle Pen is designed to facilitate the K-kit gluing operation. It has a thin needle 3.0 mm long and 0.27 mm in diameter. For use with the K-kit for TEM observation of liquid samples.
Catalog No. 50-192-8046 Supplier Electron Microscopy Sciences Supplier No. K7265
The Needle Pen is designed to facilitate the K-kit gluing operation. It has a thin needle 3.0 mm long and 0.27 mm in diameter. For use with the K-kit for TEM observation of liquid samples.