Learn More
Cooper Atkins Corporation NEEDLE PROBE 6IN FOR DIG THERM
Catalog No. NC9979071
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Dura/Needle, Technology Base: Thermocouple, Thermocouple Type: Type K, Max Tip Temperature: 500 deg. F/260 deg. C, Max Cord Temperature: 176 deg. F/80 deg. C, Response Time: 2 sec. respose in liquids, Shaft Length: 6 in./153mm
Catalog No. NC9979071 Supplier Cooper Atkins Corporation Supplier No. 50336K
This item has a minimum qty of 2 per supplier requirements.
Dura/Needle, Technology Base: Thermocouple, Thermocouple Type: Type K, Max Tip Temperature: 500 deg. F/260 deg. C, Max Cord Temperature: 176 deg. F/80 deg. C, Response Time: 2 sec. respose in liquids, Shaft Length: 6 in./153mm