Learn More
Med Vet International FlipLock Safety Needles, 18G x 1in, 50/Box
Catalog No. 502643622
Encompass_Preferred
Perhaps the most versatile and cost-effective safety needle available. EasyTouch FlipLock safety needles fit all standard Luer Lock and Slip Fit syringe barrels. Easy to Use Protective Cover Shields Needle After Use Multi-lock Safety System Latex Free
This item is not returnable. View return policy
Catalog No. 50-264-3622 Supplier Med Vet International Supplier No. 811801
Perhaps the most versatile and cost-effective safety needle available. EasyTouch FlipLock safety needles fit all standard Luer Lock and Slip Fit syringe barrels. Easy to Use Protective Cover Shields Needle After Use Multi-lock Safety System Latex Free