Learn More
Waters Corp Waters Needle Seal, Gauge 22, 10 per Package, HPLC
Catalog No. 50792697 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This needle seal is a short length of Teflon tubing designed to create a seal around a syringe needle in HPLC systems. It incorporates a stainless steel ferrule that tightens around the Teflon sleeve, ensuring a leak-proof connection.
  • Designed for HPLC systems.
  • Features a Teflon tubing seal.
  • Includes a stainless steel ferrule for a leak-proof fit.
  • Recommended replacement every 1500 to 2000 penetrations.
Catalog No. 50-792-697 Supplier Waters Corp Supplier No. 700002208
This needle seal is a short length of Teflon tubing designed to create a seal around a syringe needle in HPLC systems. It incorporates a stainless steel ferrule that tightens around the Teflon sleeve, ensuring a leak-proof connection.
  • Designed for HPLC systems.
  • Features a Teflon tubing seal.
  • Includes a stainless steel ferrule for a leak-proof fit.
  • Recommended replacement every 1500 to 2000 penetrations.