Learn More
Waters Corp Waters Corporation Needle Seat, VESPEL, ACQUITY FTN, HPLC, UPLC
Catalog No. 50466546 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This needle seat is a replacement spare part designed for use with ACQUITY Sample Managers that feature Flow Through Needle (FTN) configurations. It is compatible with both standard and Bio configurations of these systems.
  • Made of Vespel.
  • Functions as a rebuild kit component.
  • Includes 1 unit per package.
  • Can be used as a needle or sample holder.
  • Compatible with HPLC and UPLC system types.
  • Recommended for annual replacement.
  • Designed for ACQUITY Sample Managers with Flow Through Needle (FTN) configurations.
  • Applicable to standard and Bio configurations.
Catalog No. 50-466-546 Supplier Waters Corp Supplier No. 405011492
This needle seat is a replacement spare part designed for use with ACQUITY Sample Managers that feature Flow Through Needle (FTN) configurations. It is compatible with both standard and Bio configurations of these systems.
  • Made of Vespel.
  • Functions as a rebuild kit component.
  • Includes 1 unit per package.
  • Can be used as a needle or sample holder.
  • Compatible with HPLC and UPLC system types.
  • Recommended for annual replacement.
  • Designed for ACQUITY Sample Managers with Flow Through Needle (FTN) configurations.
  • Applicable to standard and Bio configurations.