Learn More
Thermo Scientific Thermostat Capillary Needle Seat 1UL
Supplier:  Thermo Scientific 6841.2472
Encompass
1 microliter needle seat capillary made of MP35N nickel-cobalt alloy for gas chromatography sample injection provides corrosion resistant durable flow path and accurate volumes via small internal diameter and tapered needle tip.
Catalog No. NC0724753