Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sciencix NEEDLE SEAT, STD PEEK 0.17MM ID
SDP

Catalog No. NC1051077
Encompass_Preferred

Needle Seat, Std., PEEK™, 0.17mm ID, Equivalent to Agilent G1329-87017

Catalog No. NC1051077 Supplier Sciencix Supplier No. 112605
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Needle Seat, Std., PEEK™, 0.17mm ID, Equivalent to Agilent G1329-87017

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.