Learn More
Trajan Scientific NEEDLE SER II NLL-5/14 5/PK
SDP
Catalog No. 502530251
Encompass_Preferred
Trajan Replaceable Luer Lock Needles: Bevel, Length Needle: 50 mm, 2 in., Needle Gauge: 14 G, Needle Point Style: Bevel, Accuracy: +/-1%, Diameter Inner: 0.062 in., 1.6 mm, Diameter Outer: 0.062 in., 2.11 mm, Diameter Inner Needle: 1.6 mm, Diameter Outer Needle: 2.1 mm
Catalog No. 50-253-0251 Supplier Trajan Scientific Supplier No. 039880
Trajan Replaceable Luer Lock Needles: Bevel, Length Needle: 50 mm, 2 in., Needle Gauge: 14 G, Needle Point Style: Bevel, Accuracy: +/-1%, Diameter Inner: 0.062 in., 1.6 mm, Diameter Outer: 0.062 in., 2.11 mm, Diameter Inner Needle: 1.6 mm, Diameter Outer Needle: 2.1 mm