Learn More
Trajan Scientific NEEDLE SER II NLL-5/18 5/PK
SDP
Catalog No. 502530252
Encompass_Preferred
Trajan Replaceable Luer Lock Needles: Bevel, Length Needle: 51 mm, 2 in., Needle Gauge: 18 G, Needle Point Style: Bevel, Diameter Inner: 0.031 in., 0.8 mm, Diameter Outer: 0.05 in., 1.27 mm, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle
Catalog No. 50-253-0252 Supplier Trajan Scientific Supplier No. 039842
Trajan Replaceable Luer Lock Needles: Bevel, Length Needle: 51 mm, 2 in., Needle Gauge: 18 G, Needle Point Style: Bevel, Diameter Inner: 0.031 in., 0.8 mm, Diameter Outer: 0.05 in., 1.27 mm, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle