Learn More
Trajan Scientific NEEDLE SER II NLL-5/19 5/PK
SDP
Catalog No. 502530254
Encompass_Preferred
Trajan Replaceable Luer Lock Needles: Bevel, Length Needle: 51 mm, 2 in., Needle Gauge: 19 G, Needle Point Style: Bevel, Diameter Inner: 0.025 in., 0.65 mm, Diameter Outer: 0.025 in., 0.64 mm, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle
Catalog No. 50-253-0254 Supplier Trajan Scientific Supplier No. 039822
Trajan Replaceable Luer Lock Needles: Bevel, Length Needle: 51 mm, 2 in., Needle Gauge: 19 G, Needle Point Style: Bevel, Diameter Inner: 0.025 in., 0.65 mm, Diameter Outer: 0.025 in., 0.64 mm, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle