Learn More
Trajan Scientific NEEDLE SER II NLL-LC 2/PK
SDP
Catalog No. 502530249
Encompass_Preferred
Trajan Replaceable Luer Lock Needles: LC, Diameter Outer: 0.027 in., 0.7 mm, Diameter Inner: 0.37 mm, 0.014 in., Needle Gauge: 22 G, Length Needle: 51 mm, 2 in., Needle Point Style: LC, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle
Catalog No. 50-253-0249 Supplier Trajan Scientific Supplier No. 039895
Add to Cart
Add to Cart
Trajan Replaceable Luer Lock Needles: LC, Diameter Outer: 0.027 in., 0.7 mm, Diameter Inner: 0.37 mm, 0.014 in., Needle Gauge: 22 G, Length Needle: 51 mm, 2 in., Needle Point Style: LC, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle