Learn More
Trajan Scientific NEEDLE SER II NLL-WLC 2/PK
SDP
Catalog No. 502530248
Encompass_Preferred
Trajan Replaceable Luer Lock Needles: LC, Diameter Outer: 0.019 in., 0.5 mm, Diameter Inner: 0.2 mm, 0.007 in., Needle Gauge: 25 G, Length Needle: 51 mm, 2 in., Needle Point Style: LC, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle
Catalog No. 50-253-0248 Supplier Trajan Scientific Supplier No. 039897
Trajan Replaceable Luer Lock Needles: LC, Diameter Outer: 0.019 in., 0.5 mm, Diameter Inner: 0.2 mm, 0.007 in., Needle Gauge: 25 G, Length Needle: 51 mm, 2 in., Needle Point Style: LC, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle