Learn More
Med Vet International Exel Needle, 20g X 3.5", Spinal, 50/Box
Catalog No. 502829186
Encompass_Preferred
This Exel International 26964 Needle is a 20G X 3.5" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is a spinal needle. It is used for entering the spinal canal or cisterna magna. This quantity is 50/Box.
Catalog No. 50-282-9186 Supplier Med Vet International Supplier No. 26964BX
This Exel International 26964 Needle is a 20G X 3.5" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is a spinal needle. It is used for entering the spinal canal or cisterna magna. This quantity is 50/Box.