Learn More
Organomation One Dozen 4in. X 16ga. Stainless Steel Needles
SDP
Catalog No. NC1336569
Encompass_Preferred
Set of one dozen luer lock needles. These needles are 4 inch by 16 gauge, with a 304 stainless steel shaft and a chrome plated brass hub.
Catalog No. NC1336569 Supplier Organomation Supplier No. NA1107
Set of one dozen luer lock needles. These needles are 4 inch by 16 gauge, with a 304 stainless steel shaft and a chrome plated brass hub.