Learn More
Practicon Dental NEEDLE STATION
SDP
Catalog No. NC1726305
Encompass
For a simple and reusable way to recap needles, try the Needle Station. It holds any size needle sheath upright for one-handed recapping as required by OSHA. Stable enough to store entire loaded syringe upright between injections. Matte finish, solid stainless steel construction. 1-1/2" tall x 2" diameter non-slip base.
Catalog No. NC1726305 Supplier Practicon Dental Supplier No. 7135939
For a simple and reusable way to recap needles, try the Needle Station. It holds any size needle sheath upright for one-handed recapping as required by OSHA. Stable enough to store entire loaded syringe upright between injections. Matte finish, solid stainless steel construction. 1-1/2" tall x 2" diameter non-slip base.