Learn More
Med Vet International Miltex Suture Needle, Size 14 With 3/8 Cirlce Taper Point, 12/Pack
Catalog No. 502832473
Encompass_Preferred
The Miltex MS-140A-14 regular surgeon's needle is a high quality suturing needle. It is a reusable Size 14 3/8 Circle Taper Point Suture Needle that comes in 12/pack.
Catalog No. 50-283-2473 Supplier Med Vet International Supplier No. MIL140A14
The Miltex MS-140A-14 regular surgeon's needle is a high quality suturing needle. It is a reusable Size 14 3/8 Circle Taper Point Suture Needle that comes in 12/pack.