Learn More
Med Vet International Miltex Suture Needle, Size 16 With 3/8 Circle Taper Point, 12/Pack
Catalog No. 502832239
Encompass_Preferred
The Miltex MS-140A-16 regular surgeon's needle is a high quality suturing needle. It is a reusable Size 16 3/8 Circle Taper Point Suture Needle that comes in 12/pack.
Catalog No. 50-283-2239 Supplier Med Vet International Supplier No. MIL140A16
The Miltex MS-140A-16 regular surgeon's needle is a high quality suturing needle. It is a reusable Size 16 3/8 Circle Taper Point Suture Needle that comes in 12/pack.