Learn More
Waters Corp Waters Corporation Needle Tip, Long, Female Luer Inlet, 20/pk
Catalog No. 50819038 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These needle tips are designed as an accessory for extraction manifolds, specifically for use with Oasis™ HLB Cartridges where stopcocks are not required. They are installed into the manifold's cover to channel cartridge effluents directly into collection vessels.
  • Long design
  • Features a female Luer inlet
  • Supplied in a pack of 20 pieces
  • Accessory for extraction manifolds
  • Used with Oasis™ HLB Cartridges
  • Channels cartridge effluents into collection vessels
Catalog No. 50-819-038 Supplier Waters Corp Supplier No. WAT200691
These needle tips are designed as an accessory for extraction manifolds, specifically for use with Oasis™ HLB Cartridges where stopcocks are not required. They are installed into the manifold's cover to channel cartridge effluents directly into collection vessels.
  • Long design
  • Features a female Luer inlet
  • Supplied in a pack of 20 pieces
  • Accessory for extraction manifolds
  • Used with Oasis™ HLB Cartridges
  • Channels cartridge effluents into collection vessels