Learn More
IDEX Health & Science Upchurch NEEDLE VALVE ASSEMBLY 1/4-28
Catalog No. NC1919689
Encompass_Preferred
Idex Micro-Metering and Micro-Splitter Valves: Needle Valves, Two Flangeless Nuts and Ferrules; P-200, P-235 Fitings, Pressure: 800, PEEK, For Use With: Chromatography Columns, Swept Volume: 7.7 uL Internal
Catalog No. NC1919689 Supplier IDEX Health & Science Upchurch Supplier No. P445
Idex Micro-Metering and Micro-Splitter Valves: Needle Valves, Two Flangeless Nuts and Ferrules; P-200, P-235 Fitings, Pressure: 800, PEEK, For Use With: Chromatography Columns, Swept Volume: 7.7 uL Internal