Learn More
Grainger Needle Valve Brass 1/4-18 5 GP
Catalog No. 502794578
Encompass_Preferred
Needle Valve Brass 1/4-18 5 GPM 2000 PSI
Catalog No. 50-279-4578 Supplier Grainger Supplier No. 1A862
Needle Valve Brass 1/4-18 5 GPM 2000 PSI