Learn More
New Jersey Fluid Systems LLC NEEDLE VALVE SS-1RS4-A
Catalog No. NC0105716
Encompass
316 Stainless Steel Swagelok Needle Valve. 1/4 in. OD Tube Fitting ends, 5,000psi @ 100F -65F to 450F, Angle Pattern PTFE Packing
Catalog No. NC0105716 Supplier New Jersey Fluid Systems LLC Supplier No. SS1RS4A
316 Stainless Steel Swagelok Needle Valve. 1/4 in. OD Tube Fitting ends, 5,000psi @ 100F -65F to 450F, Angle Pattern PTFE Packing