Learn More
Grainger Colorflow Needle Valve, In-Line, Steel, 1/2 in. NPT, 5,000 psi
Catalog No. 502794998
Encompass_Preferred
This in-line needle valve is designed for precise flow control in hydraulic and pneumatic systems, enabling flow metering in both directions or reliable shut-off. Featuring a steel body and a standard needle design, it is suitable for a wide range of industrial applications including factory automation, machine tools, and material handling.
  • Exclusive color-band reference scale on the valve stem provides an accurate visual indication of the setting
  • Manual actuation via a standard steel, zinc-plated knob
  • Operating temperature range of -40°C to +121°C
  • Maximum operating pressure of 5,000 psi
  • Maximum flow rate of 57 L/min (15 gal/min)
  • NPTF port type with 1/2 in. port size
  • Equipped with nitrile seals
  • Includes a set screw for locking the valve at a desired setting
Catalog No. 50-279-4998 Supplier Grainger Supplier No. 1A049
This in-line needle valve is designed for precise flow control in hydraulic and pneumatic systems, enabling flow metering in both directions or reliable shut-off. Featuring a steel body and a standard needle design, it is suitable for a wide range of industrial applications including factory automation, machine tools, and material handling.
  • Exclusive color-band reference scale on the valve stem provides an accurate visual indication of the setting
  • Manual actuation via a standard steel, zinc-plated knob
  • Operating temperature range of -40°C to +121°C
  • Maximum operating pressure of 5,000 psi
  • Maximum flow rate of 57 L/min (15 gal/min)
  • NPTF port type with 1/2 in. port size
  • Equipped with nitrile seals
  • Includes a set screw for locking the valve at a desired setting