Learn More
Waters Corp Waters Needle Wash Station, for 2707 Autosampler, Prevents Sample Carryover
Catalog No. 50792917 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Needle Wash Station is designed for use with the 2707 Autosampler. It effectively prevents sample carryover and contamination by performing a programmable wash routine that cleans the needle and tubing.
  • Removes and rinses sample from tubing and needle.
  • Features a programmable wash routine after each injection.
  • Aspirates wash solvent to prepare sample line and facilitate partial-loop loading.
  • Optimizes reversed-phase chromatography by matching wash solvent to initial gradient.
  • Includes wash/waste port, wash bottle, and tubing guide.
  • Compatible with water or organic wash solvents (e.g., distilled water, 80% methanol).
  • Recommends an inline degasser for improved wash solvent degassing.
  • Supports “Wash/prime needle” for tube filling and air removal.
  • Utilizes approved glass wash solvent bottle and low gas permeability tubing.
Catalog No. 50-792-917 Supplier Waters Corp Supplier No. 700003845
This Needle Wash Station is designed for use with the 2707 Autosampler. It effectively prevents sample carryover and contamination by performing a programmable wash routine that cleans the needle and tubing.
  • Removes and rinses sample from tubing and needle.
  • Features a programmable wash routine after each injection.
  • Aspirates wash solvent to prepare sample line and facilitate partial-loop loading.
  • Optimizes reversed-phase chromatography by matching wash solvent to initial gradient.
  • Includes wash/waste port, wash bottle, and tubing guide.
  • Compatible with water or organic wash solvents (e.g., distilled water, 80% methanol).
  • Recommends an inline degasser for improved wash solvent degassing.
  • Supports “Wash/prime needle” for tube filling and air removal.
  • Utilizes approved glass wash solvent bottle and low gas permeability tubing.