Learn More
Med Vet International Exel Needle, 14G X 1in, Hypodermic, Vet Use, 100/BX, 26450
Catalog No. 502092503
Encompass_Preferred
This Exel International 26450 Needle is a 14G X 1' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2503 Supplier Med Vet International Supplier No. 26450
 Restricted Item
Add to Cart
Add to Cart
This Exel International 26450 Needle is a 14G X 1' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX