Learn More
Med Vet International Exel Needle, 16G X 1in, Hypodermic, Vet Use, 100/BX, 26457
Catalog No. 502092506
Encompass_Preferred
This Exel International 26457 Needle is a 16G X 1' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2506 Supplier Med Vet International Supplier No. 26457
This Exel International 26457 Needle is a 16G X 1' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX