Learn More
Med Vet International Exel Needle, 16G X 3/4in, Hypodermic, Vet Use, 100/BX, 26456
Catalog No. 502092508
Encompass_Preferred
This Exel International 26456 Needle is a 16G X 3/4' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-209-2508 Supplier Med Vet International Supplier No. 26456
This Exel International 26456 Needle is a 16G X 3/4' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX